missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Healthcare™ PROTOCOL™ 10% Buffered Formalin-Filled Containers
Specifically engineered for storage of histological tissue specimens
Supplier: Fisher Healthcare™ 426796
Description
- PET
- Crystal-clear, non-autoclavable
- Flexible containers are resistant to alkalis and weak acids
- Prefilled containers are supplied with silk-screened warning label
Chemical Identifiers
| PET |
Specifications
| PET | |
| 8 oz. | |
| 240 mL, pre-filled 120 mL |