missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ 10% Buffered Formalin-Filled Containers

Catalog No. p-181197
Change view
Click to view available options
Capacity (Metric):
240 mL, pre-filled 120 mL
480 mL
Capacity (English):
8 oz.
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Capacity (English)
23426797 480 mL 16 oz.
23426796 240 mL, pre-filled 120 mL 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23426797 Supplier Fisher Healthcare™ Supplier No. 426797
Only null left

Specifically engineered for storage of histological tissue specimens

  • PET
  • Crystal-clear, non-autoclavable
  • Flexible containers are resistant to alkalis and weak acids
  • Prefilled containers are supplied with silk-screened warning label

Specifications

Material PET
Capacity (English) 16 oz.
Capacity (Metric) 480 mL