missing translation for 'onlineSavingsMsg'
Learn More

Fisher Container™ Precision Zip-Clean Pouches™

Easy open and easy zip closure.

$1,322.38 - $2,376.39

Specifications

Closure Type Zip Closure
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
19351161
Encompass
View Documents Promotion Details
Fisher Container™
91088P
10 x 12 in. (26 x 31cm) 1000
Case of 1000 Each for $1,322.38
Only null left
 
19351163
Encompass
View Documents Promotion Details
Fisher Container™
91090P
18 x 24 in. (46 x 61cm) 400
Case of 400 Each for $1,507.45
Only null left
 
19351162
Encompass
View Documents Promotion Details
Fisher Container™
STCK0072001
12 x 15 in. (31 x 38cm) 1000
Case of 1000 Each for $1,865.73
Only null left
 
19351160
Encompass
View Documents Promotion Details
Fisher Container™
STCK0071001
8 x 10 in. (21 x 26cm) 2000
Case of 2000 Each for $2,376.39
Only null left
 
19351158
Encompass
View Documents Promotion Details
Fisher Container™
STCK0049001
4 x 6 in. (11 x 16cm) 2000
Case of 2000 Each for $1,867.25
Only null left
 
Description

Description

  • Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications
  • Cleanroom certified
  • Sealed in a double-lined carton
  • Barrier properties

Specifications

Specifications

Zip Closure
Videos
SDS
Documents

Documents