Learn More
Molecular Probes™ FISH Tag™ DNA Kits with Alexa Fluor™ Dyes
FISH Tag DNA kits provide a complete solution for fluorescence in situ hybridization (FISH) applications. Choose from kits using Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, or multicolor dyes for probe synthesis, purification, and labeling of specimens for imaging.
Supplier: Molecular Probes™ F32948
Description
Choose from three, single-color FISH Tag DNA kits with Alexa Fluor dye options (green, orange, or red). The FISH DNA multicolor kit contains four spectrally distinct dyes (green, orange, red, and deep-red) to allow simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and in situ analysis of mitochondrial DNA localization and content. All kits include 16 reagents and come with a comprehensive, detailed protocol to help ensure your success.
FISH Tag™ DNA Kits provide a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. Choose from FISH Tag™ DNA Kits in three Alexa Fluor™ colors or get the FISH Tag™ DNA Multicolor Kit (Cat. No. F32951), which contains four spectrally distinct dyes to allow simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and for in situ analysis of mitochondrial DNA localization and content. All kits include 16 reagents and come with a comprehensive, detailed protocol to help ensure your success.
FISH Tag™ Kit Specifications:
- Dye (Ex/Em): Alexa Fluor™ 594 (590/615 nm)
- Comprehensive kit includes 16 reagents
- Comes with detailed protocol to help ensure your success
Complete Workflow Solution for FISH
FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. PureLink™ nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade™ Gold antifade reagent is added to preserve the fluorescent signal during imaging.
More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Chemical Identifiers
| Store in freezer (-5 to -30°C) and protect from light. | |
| Kit | |
| DNA, Tissue (FFPE), Tissue (CRYO), Cell Culture Cells | |
| FISH Tag, fluorescent microscopy, DNA In situ hybridization | |
| Alexa Fluor, FISH Tag | |
| 1 Kit | |
| Not Target-Specific |
| Fluorescence | |
| Probes (Labeled DNA), cDNA (Labeled) | |
| DNA (General), cDNA | |
| 10 Reactions | |
| DNA In Situ Hybridization Assay Kit | |
| DNA | |
| Dry Ice |
Specifications
| Store in freezer (-5 to -30°C) and protect from light. | |
| Fluorescence | |
| Alexa Fluor 555 | |
| DNA, Tissue (FFPE), Tissue (CRYO), Cell Culture Cells | |
| 555/565 nm | |
| 10 Reactions | |
| DNA In Situ Hybridization Assay Kit | |
| DNA | |
| Dry Ice |
| Orange | |
| Kit | |
| Probes (Labeled DNA), cDNA (Labeled) | |
| DNA (General), cDNA | |
| FISH Tag, fluorescent microscopy, DNA In situ hybridization | |
| Alexa Fluor, FISH Tag | |
| 1 Kit | |
| Not Target-Specific |
For Research Use Only. Not for use in diagnostic procedures.