missing translation for 'onlineSavingsMsg'
Learn More

Fipronil 97.0+%, TCI America™

Catalog No. F08225G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08225G Supplier TCI America Supplier No. F08225G

Specifications

CAS 120068-37-3
Melting Point 204°C
Color White-Yellow
Molecular Formula C12H4Cl2F6N4OS
Quantity 5 g
MDL Number MFCD00867812
UN Number 2811
Synonym fipronil, fipronil iso, 5-amino-1-2,6-dichloro-4-trifluoromethyl phenyl-4-trifluoromethyl sulfinyl-1h-pyrazole-3-carbonitrile, 5-amino-1-2,6-dichloro-4-trifluoromethyl phenyl-4-trifluoromethylsulfinyl pyrazole-3-carbonitrile, 5-amino-3-cyano-1-2,6-dichloro-4-trifluoromethylphenyl-4-trifluoromethylsulfinylpyrazole, dsstox_cid_14609, dsstox_rid_79176, dsstox_gsid_34609, 1h-pyrazole-3-carbonitrile, 5-amino-1-2,6-dichloro-4-trifluoromethyl phenyl-4-trifluoromethyl sulfinyl
InChI Key ZOCSXAVNDGMNBV-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Cl)N2C(=C(C(=N2)C#N)S(=O)C(F)(F)F)N)Cl)C(F)(F)F
IUPAC Name 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethylsulfinyl)pyrazole-3-carbonitrile
Molecular Weight (g/mol) 437.141
PubChem CID 3352
ChEBI CHEBI:83394
Formula Weight 437.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fipronil
Show More Show Less
EINECSNumber (5)-6414
RTECSNumber UQ4430250
TSCA No
Recommended Storage Refrigerator