Learn More
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipette PROMO
Catalog No. 14386319TI
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
0.5 to 5 μL
0.5 to 5 mL
1 to 10 μL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
Color:
Blue
Green
Pink
Red
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14386319TI 20 to 200 μL Yellow
14386317TI 2 to 20 μL Yellow
14386312TI 0.2 to 2 μL Pink
14386315TI 1 to 10 μL Yellow
14386314TI 1 to 10 μL Pink
14386313TI 0.5 to 5 μL Pink
14386321TI 0.5 to 5 mL Green
14386322TI 1 to 10 mL Red
14386318TI 10 to 100 μL Yellow
14386320TI 100 to 1000 μL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14386319TI Supplier Thermo Scientific™ Supplier No. P4642080
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of these fully autoclavable pipettes.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
Accuracy ±1.8 to 0.6%
Compatible Tips Finntip 250, Filter 200, Flex 200, Flex Filter 200, 200 Ext, Filter 200 Ext, Wide 250
Autoclavable Autoclavable
Color Yellow
Model F2
Number of Channels 1
Product Line Finnpipette™ F2
Type Variable Volume Pipette
Volume (Metric) 20 to 200 μL
Increments 0.2 μL
Precision 0.7 to 0.2%
Quantity 1 Ea.
Show More Show Less