Learn More
Thermo Scientific™ Finnpipette™ F2 Multichannel Pipettes
GREENER_CHOICE
Catalog No. p-4530175
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
12
16
8
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
14386359 1 to 10 μL 8
14386360 5 to 50 μL 8
14386361 10 to 100 μL 8
14386362 30 to 300 μL 8
14386363 1 to 10 μL 12
14386364 5 to 50 μL 12
14386365 10 to 100 μL 12
14386366 30 to 300 μL 12
14386367 1 to 10 μL 16
14386368 5 to 50 μL 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14386359 Supplier Thermo Scientific™ Supplier No. 4662000
Only null left
Use this multichannel pipette for highly demanding applications where robustness and durability are essential.
Use the Thermo Scientific™ Finnpipette™ F2 Multichannel Pipette for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, this pipette is built with PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable and available in 8-, 12- and 16-channel models with a selection of volume ranges. In the low volume models, the super blow-out function ensures accurate dispensing for even the lowest volumes.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F2 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country
Specifications
Compatible Tips Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Description Finnpipette™ F2
Autoclavable Autoclavable
Color Pink
Number of Channels 8
Product Line Finnpipette™ F2
Type Pipette
Volume (Metric) 1 to 10 μL
Increments 0.02μL