Learn More
Thermo Scientific™ Finnpipette™ F2 Multichannel Pipette PROMO
Catalog No. 14386366TI
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
12
8
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
14386366TI 30 to 300 μL 12
14386361TI 10 to 100 μL 8
14386359TI 1 to 10 μL 8
14386360TI 5 to 50 μL 8
14386362TI 30 to 300 μL 8
14386365TI 10 to 100 μL 12
14386363TI 1 to 10 μL 12
14386364TI 5 to 50 μL 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14386366TI Supplier Thermo Scientific™ Supplier No. P4662070
Only null left
Use this multichannel pipette for highly demanding applications where robustness and durability are essential.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
Compatible Tips Finntip 300, Filter 300, Flex 300, Flex Filter 300
Autoclavable Autoclavable
Color Orange
Model F2
Number of Channels 12
Product Line Finnpipette™ F2
Type Pipette
Volume (Metric) 30 to 300 μL
Increments 1 μL
Precision 2 to 0.3%
Quantity 1 Ea.
Show More Show Less