Learn More
Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes
GREENER_CHOICE
Catalog No. 14386300N
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
0.5 to 5 μL
0.5 to 5 mL
1 to 10 μL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 50 μL
Color:
Blue
Green
Orange
Pink
Red
Turquoise
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14386300N 0.2 to 2 μL Pink
14386301N 0.5 to 5 μL Pink
14386302N 1 to 10 μL Pink
14386303N 1 to 10 μL Yellow
14386304N 2 to 20 μL Turquoise
14386305N 2 to 20 μL Yellow
14386306N 10 to 100 μL Yellow
14386307N 20 to 200 μL Yellow
14386308N 30 to 300 μL Orange
14386309N 100 to 1000 μL Blue
14386310N 0.5 to 5 mL Green
14386311N 1 to 10 mL Red
14386292N 5 to 50 μL Turquoise
14386293N 5 to 50 μL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 14386300N Supplier Thermo Scientific™ Supplier No. 4641010N
Maximize user comfort and reduce strain with the F1 system, which delivers performance with precision, all in a lightweight design. It's the ideal system for applications using expensive reagents and valuable samples.
Enjoy the enhanced volume adjustment on Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes, which is now silent and effortless with textured adjustment knob for a firmer feel when setting volumes. The adjustable finger rest and lightweight design maximize comfort without sacrificing performance
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Compatible Tips Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Color Pink
Model Finnpipette F1
Number of Channels 1
Type Variable Volume Pipette
Volume (Metric) 0.2 to 2 μL
Increments 0.002μL