Learn More
Thermo Scientific™ Finnpipette™ F1 Multichannel Pipettes
GREENER_CHOICE
Catalog No. 14386349N
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
12
16
8
Color:
Orange
Pink
Purple
Turquoise
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Color
14386349N 1 to 10 μL 8 Pink
14386350N 5 to 50 μL 8 Yellow
14386351N 10 to 100 μL 8 Yellow
14386352N 30 to 300 μL 8 Orange
14386353N 1 to 10 μL 12 Pink
14386354N 5 to 50 μL 12 Yellow
14386355N 10 to 100 μL 12 Yellow
14386356N 30 to 300 μL 12 Orange
14386357N 1 to 10 μL 16 Purple
14386358N 5 to 50 μL 16 Turquoise
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14386349N Supplier Thermo Scientific™ Supplier No. 4661000N
Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.
Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It's the ideal system for applications using expensive reagents and valuable samples.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Compatible Tips Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Description Finnpipette F1
Color Pink
Number of Channels 8
Volume (Metric) 1 to 10 μL
Increments 0.02μL