Learn More
Thermo Scientific™ Finnpipette™ Dispenser
GREENER_CHOICE
Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.
Supplier:  Thermo Scientific™ 4421130
 View more versions of this product
Catalog No. 14487201
Only null left
Description
Description
Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.
Features:
  • Excellent chemical and thermal resistance
  • Fully autoclavable†
  • Exact self-locking system in the volume setting
  • Dispensing head cap prevents dripping
  • Ergonomic design prevents work fatigue
  • Threaded suction hose for bubble-free suction
Note:
Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.
Specifications
Chemical Identifiers
Analog Bottletop Dispenser
Individual calibration certificate with serial number
1
Specifications
Analog Bottletop Dispenser
Range: 0.4-2.0mL, ±12.0µL
0.4 to 2.0 mL
Individual calibration certificate with serial number
1
4 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Note: Not for use on hot plates.