missing translation for 'onlineSavingsMsg'
Learn More

Fingolimod Hydrochloride 98.0+%, TCI America™

Catalog No. p-7130028
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1018200MG 200 mg
F10181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1018200MG Supplier TCI America Supplier No. F1018200MG
Only null left

Chemical Identifiers

CAS 162359-56-0
Molecular Formula C19H34ClNO2
Molecular Weight (g/mol) 343.936
MDL Number MFCD00939512
InChI Key SWZTYAVBMYWFGS-UHFFFAOYSA-N
Synonym 2-Amino-2-[2-(4-n-octylphenyl)ethyl]propane-1,3-diol Hydrochloride, FTY 720
PubChem CID 107969
ChEBI CHEBI:63112
IUPAC Name 2-amino-2-[2-(4-octylphenyl)ethyl]propane-1,3-diol;hydrochloride
SMILES CCCCCCCCC1=CC=C(C=C1)CCC(CO)(CO)N.Cl

Specifications

Melting Point 110°C
Color White
Quantity 200 mg
Formula Weight 343.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fingolimod Hydrochloride
RTECSNumber TY3130000
TSCA No
Recommended Storage Freezer (-20 °C)