Learn More
Thermo Scientific™ Finger Tite Ferrules
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
$245.67 - $279.14
Description
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific Finger Tite ferrules are the next generation of the SilTite™ connection system.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- No tools required
Specifications
| Ferrule |