Learn More
Corning™ PYREX™ Filling Bells
For aseptically filling culture tubes and bottles
$177.98 - $355.12
| Catalog Number | Mfr. No. | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
|
14983A
|
Corning™
3960S |
7 mm | 70 mm |
Case for $177.98
|
|
||||
1185007
![]() |
Corning™
396550 |
9 mm | 105 mm |
Case of 2 Each for $355.12
|
|
||||
14983B
![]() |
Corning™
3960L |
11 mm | 85 mm |
Case for $203.30
|
|
||||
Description
For aseptic filling of vessels. Rubber tubing is attached to the top of the tubulation while the receiving vessel is placed inside the bell.
Specifications
| Borosilicate Glass |