missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Filaggrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5116911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116911 Supplier Invitrogen™ Supplier No. PA5116911
Only null left

Rabbit Polyclonal Antibody

Positive Samples: A375, A431, HepG2, TE-1, SGC7901, mouse stomach, mouse liver, rat liver Immunogen sequence: MSTLLENIFA IINLFKQYSK KDKNTDTLSK KELKELLEKE FRQILKNPDD PDMVDVFMDH LDIDHNKKID FTEFLLMVFK LAQAYYESTR KE.

Filaggrin is expressed only in well differentiated keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.

Specifications

Antigen Filaggrin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene FLG
Gene Accession No. P11088, P20930
Gene Alias ATOD2; AW107830; epidermal filaggrin; filaggrin; Filaggrin (profilaggrin); FLG; ft; Pflg
Gene Symbols FLG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-92 of human FLG (NP_0020071).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14246, 2312, 24641
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less