missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Field's Stain B

CAS: 17372-87-1 | C20H6Br4Na2O5 | 691.86 g/mol

$57.45 - $181.05

Chemical Identifiers

CAS 17372-87-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1857809
View Documents Promotion Details
Thermo Scientific Chemicals
A1857809
10g
Each for $57.45
Only null left
 
AAA1857818
View Documents Promotion Details
Thermo Scientific Chemicals
A1857818
50g
Each for $181.05
Only null left
 
Description

Description

Used to stain blood smears to identify malarial parasites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17372-87-1

Specifications

17372-87-1
SEACYXSIPDVVMV-UHFFFAOYSA-L
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
691.86
C20H6Br4Na2O5
Stain
disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only