missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibulin 2 Monoclonal Antibody (7H4F6/3B9A11)
GREENER_CHOICE

Catalog No. PIMA515390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515390 Supplier Invitrogen™ Supplier No. MA515390
Only null left

Mouse Monoclonal Antibody

MA5-15390 targets FBLN2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15390 immunogen is purified recombinant fragment of FBLN2 (aa180-440) expressed in E. Coli.

This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen Fibulin 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7H4F6/3B9A11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene FBLN2
Gene Accession No. P98095
Gene Alias 5730577E14Rik; FBLN2; FIBL2; FIBL-2; fibulin 2; Fibulin2; fibulin-2
Gene Symbols FBLN2
Host Species Mouse
Immunogen Purified recombinant fragment of FBLN2 (aa180-440) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less