missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fibronectin Human Protein, Plasma

Catalog No. 33016015
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33016015 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33016015 Supplier Gibco™ Supplier No. 33016015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fibronectin Human Protein, Plasma is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules.

Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules. Human Plasma Fibronectin is isolated from human plasma by affinity chromtography. Fibronectin has a molecular weight of 440,000 and is made up of two 220,000 dalton polypeptides. Human Fibronectin is filtered (0,2UI) and supplied lyophilized.

Performance and Quality Testing

The purity is determined by SDS-PAGE. Bovine and human fibronectins are assayed for their ability to support cell attachment and spreading, and screened to confirm absence of mycoplasma or bacterial contamination.

Specifications

For Use With (Application) Cell Culture
Quantity 5 mg
Packaging Glass Bottle
Gene Alias Fibronectin Human, Plasma
Species Human
Recombinant Native
Sterility Sterile
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Fibronectin
Research Category Extracellular Matrix & Cell Adhesion, Stem Cell Research
Color White
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.