missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fibronectin-Binding Protein

For research use

$329.04 - $579.74

Specifications

Formulation Lyophilized powder
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Fibronectin-Binding Protein
Recombinant Native
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66903EXD
View Documents
Thermo Scientific Chemicals
J66903EXD
0.5mg
Each for $329.04
Only null left
 
AAJ66903MCR
View Documents
Thermo Scientific Chemicals
J66903MCR
1mg
Each for $579.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lyophilized powder
RUO
Native
-30° to -10°C
Fibronectin-Binding Protein
Solid
Videos
SDS
Documents

Documents

RUO – Research Use Only