missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fibronectin-Binding Protein

Catalog No. AAJ66903MCR
Change view
Click to view available options
Quantity:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66903MCR 1mg
AAJ66903EXD 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66903MCR Supplier Thermo Scientific Chemicals Supplier No. J66903MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For research use

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Formulation Lyophilized powder
Quantity 1mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Fibronectin-Binding Protein
Recombinant Native
Form Solid

RUO – Research Use Only