missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen gamma Monoclonal Antibody (K24024_13F12), Invitrogen™
Mouse Monoclonal Antibody
$1740.58 - $2926.56
Specifications
| Antigen | Fibrinogen gamma |
|---|---|
| Clone | K24024_13F12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: KVDKDLQSLE DILHQVENKT SEVKQLIKAI QLTYNPDESS KPNMIDAATL KSRKMLEEIM KYEASILTHD SSIRYLQEIY NSNNQKIVNL KEKVAQLEAQ
Fibrinogen gamma Monoclonal antibody specifically detects Fibrinogen gamma in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| Fibrinogen gamma | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
| Recombinant human FGG (immunogen range is 61-160). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Fgg |
| K24024_13F12 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P02679 | |
| 2266 | |
| IgG1 | |
| Protein A | |
| Antibody |