missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen alpha chain Recombinant Rabbit Monoclonal Antibody (23GB5400)
GREENER_CHOICE

Catalog No. MA552850
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552850 100 μL
MA552851 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552850 Supplier Invitrogen™ Supplier No. MA552850
Only null left

Rabbit Recombinant Monoclonal Antibody

Fibrinogen alpha chain Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.

Specifications

Antigen Fibrinogen alpha chain
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5400
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Fga
Gene Accession No. E9PV24, P02671, P06399
Gene Alias Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogenα chain; Fibrinopeptide A
Gene Symbols Fga
Host Species Rabbit
Immunogen A synthesized peptide derived from human Fibrinogen alpha chain (750-800AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14161, 2243, 361969
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less