missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrinogen alpha chain Monoclonal Antibody (KT9)

Rat Monoclonal Antibody
Supplier: Invitrogen™ MA549571
Description
This product was used in both Indirect and Sandwich ELISA.
Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.Chemical Identifiers
| Fibrinogen alpha chain | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P02671 | |
| 2243 | |
| Fibrinogen purified from human blood containing a,β and ? chains. | |
| Antibody |
| KT9 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human | |
| Fga | |
| Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogenα chain; Fibrinopeptide A | |
| Fga | |
| Primary | |
| Protein G |
Specifications
| Fibrinogen alpha chain | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P02671 | |
| Fga | |
| Fibrinogen purified from human blood containing a,β and ? chains. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| KT9 | |
| Unconjugated | |
| Fga | |
| Ac1873; ENSMUSG00000059807; Fba5e; Fga; Fib; Fib2; Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogenα chain; Fibrinopeptide A | |
| Rat | |
| Protein G | |
| RUO | |
| 2243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |