missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrillarin Recombinant Rabbit Monoclonal Antibody (23GB5735)

Catalog No. MA552901
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552901 20 μL
MA552900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552901 Supplier Invitrogen™ Supplier No. MA552901
Only null left

Rabbit Recombinant Monoclonal Antibody

Fibrillarin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).

Specifications

Antigen Fibrillarin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5735
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene fbl
Gene Accession No. P22087, P22509, P35550
Gene Alias 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7
Gene Symbols fbl
Host Species Rabbit
Immunogen A synthesized peptide derived from human Fibrillarin (270-321AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14113, 2091, 292747
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less