missing translation for 'onlineSavingsMsg'
Learn More

Youngstown Barrel & Drum Fiber Drums - PROMO

Youngstown Barrel & Drum fiber drums are constructed of sturdy fiberboard and are an excellent solution for the containment and storage of dry or solid materials.

$83.11

Specifications

For Use With (Application) For transport of dry, nonreactive solid materials only
Material Fiber
Head Style Open
View More Specs

Products 1
Catalog Number Mfr. No. Certifications/Compliance Diameter (English) Inner Height (English) Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. Certifications/Compliance Diameter (English) Inner Height (English) Weight (English) Price Quantity & Availability  
18120244PMO
View Documents Promotion Details
Youngstown Barrel & Drum
1987PMO
UN 1G/Y100/S 21.5 in. 35 in. 17.4 lb.
Each for $83.11
Only null left
 
Description

Description

  • Open-head drums qualify for transport of dry, nonreactive solid materials only
  • Available with all-fiber construction (fiber covers) or with steel chime bands and metal cover
Specifications

Specifications

For transport of dry, nonreactive solid materials only
Fiber
Open
Videos
SDS
Documents

Documents