missing translation for 'onlineSavingsMsg'
Learn More

FHL2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Numéro de catalogue. 501557389
Encompass
Modifier l'affichage
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
501557389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 501557389 Fournisseur Bethyl Laboratories Inc Code fournisseur A300332A

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

LIM proteins contain a highly conserved double zinc finger motif called the LIM domain.

Spécifications

Antigen FHL2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene FHL2
Gene Accession No. O70433, Q14192
Gene Alias Four-and-a-half LIM domains 2
Gene Symbols FHL2
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14200, 2274
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats