missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGF5 Monoclonal Antibody (K1E008_4E10), Invitrogen™

Catalog No. MA556205
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556205 200 μL
MA555825 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556205 Supplier Invitrogen™ Supplier No. MA556205

Mouse Monoclonal Antibody

Sequence of this protein is as follows: YNTYASAIHR TEKTGREWYV ALNKRGKAKR GCSPRVKPQH ISTHFLPRFK QSEQPELSFT VTVPEKKKPP SPIKPKIPLS APRKNTNSVK

Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.

Specifications

Antigen FGF5
Applications Western Blot
Classification Monoclonal
Clone K1E008_4E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene FGF5
Gene Accession No. P12034
Gene Alias angora; FGF; FGF5; FGF-5; Fibroblast growth factor; Fibroblast growth factor 5; fibroblast growth factor FGF-5; go; HBGF-5; Heparin-binding growth factor 5; Smag-82; TCMGLY
Gene Symbols FGF5
Host Species Mouse
Immunogen Recombinant human FGF5 (immunogen range is 171-260).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less