missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGF20 Monoclonal Antibody (K24028_13C3), Invitrogen™

Catalog No. MA556188
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556188 200 μL
MA555808 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556188 Supplier Invitrogen™ Supplier No. MA556188
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: LGGLEGLGQQ VGSHFLLPPA GERPPLLGER RSAAERSARG GPGAAQLAHL HGILRRRQLY CRTGFHLQIL PDGSVQGTRQ DHSLFGILEF ISVAVGLVSI RGVDSGLYLG MNDKGELYGS EKLTSECIFR EQFEENWYNT YSSNIYKHGD TGRRYFVALN KDGTPRDGAR SKRHQKFTHF LPRPVDPERV PELYKDLLMY

FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.

Specifications

Antigen FGF20
Applications Western Blot
Classification Monoclonal
Clone K24028_13C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene FGF20
Gene Accession No. Q9NP95
Gene Alias FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2
Gene Symbols FGF20
Host Species Mouse
Immunogen Recombinant human FGF20 (immunogen range is 11-210).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26281
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less