missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF1 Monoclonal Antibody (C14)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566054
Description
FGF1 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF
FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.Specifications
| FGF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P05230, P20002 | |
| FGF1 | |
| Recombinant Fibroblast Growth Factor 1, Acidic (FGF1) (Met1-Asp155). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C14 | |
| Unconjugated | |
| FGF1 | |
| acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2246, 397497 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |