missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine Hydrochloride 98.0+%, TCI America™

Catalog No. F06981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06981G 1 g
F06985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06981G Supplier TCI America Supplier No. F06981G

Chemical Identifiers

CAS 153439-40-8
Molecular Formula C32H40ClNO4
Molecular Weight (g/mol) 538.13
MDL Number MFCD00865710
InChI Key RRJFVPUCXDGFJB-UHFFFAOYNA-N
Synonym fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride
PubChem CID 63002
IUPAC Name hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
SMILES [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 538.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fexofenadine Hydrochloride
RTECSNumber CY1633375
TSCA No
Recommended Storage Freezer