missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine HCl Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013227
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013227 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013227 Supplier Merck Emd Millipore Supplier No. PHR16851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fexofenadine HCl belongs to the class of drugs known as antihistamines. It acts by selective blockade of H1-receptors and is generally used in the treatment of seasonal allergic rhinitis.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 153439-40-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C32H39 NO4 · HCl
MDL Number MFCD00865710
Synonym Fexofenadine hydrochloride; Fexofenidine hydrochloride; MDL 16455 hydrochloride; Terfenidine carboxylate hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 538.12
Grade Certified Reference Material