missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fetuin B Monoclonal Antibody (K49014_16D1), Invitrogen™

Catalog No. MA556153
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556153 200 μL
MA555773 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556153 Supplier Invitrogen™ Supplier No. MA556153
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: VNQKPTNLPK VEESQQKNTP PTDSPSKAGP RGSVQYLPDL DDKNSQEKGP QEAFPVHLDL TTNPQGETLD ISFLFLEPME

The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.

Specifications

Antigen Fetuin B
Applications Western Blot
Classification Monoclonal
Clone K49014_16D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene FETUB
Gene Accession No. Q9UGM5
Gene Alias 16G2; 2310011O17Rik; AI255764; D17980; Fet; FETUB; fetuin B; fetuin beta; fetuin phosphoprotein; fetuin-B; Fetuin-like protein IRL685; Gugu; IRL685; Pp63
Gene Symbols FETUB
Host Species Mouse
Immunogen Recombinant human FETUB (immunogen range is 271-350).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less