missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fetuin A Monoclonal Antibody (OTI2H2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525949

Catalog No. PIMA525949

Only null left

Description

Description

Fetuin A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
Specifications

Chemical Identifiers

Fetuin A
1.00 mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P02765
197
Full length human recombinant protein of AHSG produced in HEK293T cell.
Antibody
OTI2H2
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgG1
Human
AHSG
59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
AHSG
Primary
Affinity chromatography

Specifications

Fetuin A
Monoclonal
1.00 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P02765
AHSG
Full length human recombinant protein of AHSG produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI2H2
Unconjugated
AHSG
59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Mouse
Affinity chromatography
RUO
197
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions