missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fetuin A Monoclonal Antibody (K56007_7D7), Invitrogen™

Catalog No. MA555745
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555745 50 μL
MA556125 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. MA555745 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' MA555745
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: DVRKVCQDCP LLAPLNDTRV VHAAKAALAA FNAQNNGSNF QLEEISRAQL VPLPPSTYVE FTVSGTDCVA KEATEAAKCN LLAEKQYGFC KATLSEKLGG AEVAVTCMVF QTQPVSSQPQ PEGANEAVPT PVVDPDAPPS

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.

Specifications

Antigen Fetuin A
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K56007_7D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene AHSG
Gene Accession No. P02765
Gene Alias 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Gene Symbols AHSG
Host Species Mouse
Immunogen Recombinant human AHSG (immunogen range is 141-280).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less