missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fetal Hemoglobin Monoclonal Antibody (HBF-1), FITC, Invitrogen™

Catalog No. MHFH014
Change view
Click to view available options
Quantity:
500 μL
2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHFH014 2 mL
501137518 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MHFH014 Supplier Invitrogen™ Supplier No. MHFH014
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility. This Fetal Hemoglobin monoclonal antibody specifically recognizes fetal hemoglobin found in fetal erythrocytes.

Hemorrhage of cells from an Rh+ fetus into the circulation of an Rh- mother may result in the formation of Rh-reactive antibodies in the mother. Rh hemolytic anemia in a subsequent Rh+ fetus may result from placental transfer of antibodies formed in the mother to the fetus. Although the volume of fetal erythrocytes found in the maternal circulation during pregnancy and immediately post-partum is reported to be small and of uncertain clinical significance in many cases, substantial hemorrhage may result from a number of causes including fetal or maternal trauma and placental defects. Erythrocytes containing fetal hemoglobin may be found in individuals of any age, but with lower amounts of fetal hemoglobin compared to fetal red cells. These cells have been termed F cells. High levels of F cells may also exist in adults with a heterogeneous group of genetic disorders of uncertain etiology, referred to as Hereditary Persistence of Fetal Hemoglobin. Other clinical conditions causing significant levels of anemia may also result in elevated levels of F cells. Several clinical conditions have been described with increased levels of F cells. These conditions include hereditary anemic diseases such as sickle cell anemia and thalassemia major.

Specifications

Antigen Fetal Hemoglobin, Fetal Hemoglobin (HbF)
Applications Flow Cytometry
Classification Monoclonal
Clone HBF-1
Conjugate FITC
Formulation PBS with <10% sucrose, BSA and 0.1% sodium azide
Gene HBG1
Gene Accession No. P69891
Gene Alias A-gamma globin; alpha-1 globin; gamma A hemoglobin; gamma globin; gamma-1-globin; Hb F Agamma; HBG1; HBGA; HBGR; HBG-T2; hemoglobin alpha 1; Hemoglobin gamma-1 chain; hemoglobin gamma-a chain; hemoglobin subunit gamma 1; hemoglobin subunit gamma-1; hemoglobin, gamma A; hemoglobin, gamma, regulator of; HSGGL1; PRO2979
Gene Symbols HBG1
Host Species Mouse
Immunogen Human Fetal Hemoglobin (HbF).
Purification Method Purified
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less