missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, ultra-low IgG, US origin
GREENER_CHOICE

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

$229.00 - $1,810.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Country of Origin United States
Form Liquid
Product Type Fetal Bovine Serum
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
A3381901
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
A3381901
50 mL One Shot
Each for $229.00
Only null left
 
16250078
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
16250078
500 mL Bottle
Each for $1,810.00
Only null left
 
Description

Description

Gibco Fetal bovine sera include a broad range of specialty sera, earning the trust of researchers with consistent quality and award-winning support that helps meet your unique research requirements. Ultra-low IgG FBS enhances the quality and accuracy of experimental results by minimizing the potential negative effects of IgG (immunoglobulin G). IgG can interfere with immunological and viral assays or experiments involving antibodies, and some cell types are sensitive to the presence of IgG.

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

IgG levels are less than 5 μg/mL, and the BVD titer is low or not detectable.

This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Fetal Bovine Serum
Fetal
Liquid
Frozen
Videos
SDS
Documents

Documents

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.