missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Fetal Bovine Serum, Regular, USDA Approved Origin

Description
- Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
- True pooled and triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
Specifications
Specifications
| Content And Storage | -40°C to -10°C |
| Species | Bovine |
| Country of Origin | United States |
| Form | Liquid |
| Quantity | 50 mL |
| Serum Treatment | Heat-inactivated |
| Sterility | Sterile |
| Source | USDA Origin |
| Heat Inactivated | Yes |