Learn More
Gibco™ Fetal Bovine Serum, Premium Plus

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk.
Supplier: Gibco™ A5669502
Description
Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk. It is ideal for use in biomanufacturing processes and diagnostic cell culture research requiring the most stringent quality profile, including an extensive biochemical and hormonal profile. Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Sera category: Premium Plus
- Use for a broad range of cell types, especially sensitive cell lines
- Our most characterized serum with the lowest endotoxin and hemoglobin levels
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤20 mg/dL
- Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco serum delivers
- ISO 13485-certified; processed in FDA-registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Chemical Identifiers
| Fetal | |
| Frozen | |
| Sterile | |
| *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. |
| Refer to CoA for specific origin* | |
| Fetal Bovine Serum | |
| Triple-filtered, 0.1 μm |
Specifications
| Store at ≤-10°C. | |
| Fetal | |
| Frozen | |
| Fetal Bovine Serum | |
| Standard (Sterile-filtered) | |
| Triple-filtered, 0.1 μm | |
| Triple-filtered, 0.1 μm | |
| ≤5 EU/mL | |
| Yes | |
| Dry Ice | |
| No |
| Cattle/Bovine | |
| Refer to CoA for specific origin* | |
| One Shot | |
| 10 x 50 mL | |
| Sterile | |
| Premium Plus | |
| Up to 70 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling | |
| *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. | |
| ≤20 mg/dL | |
| USDA Safety Testing Performed for Mexican and Australian origins |
Safety and Handling
ShelfLife : 5 Years
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.