missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, mesenchymal stem cell-qualified, USDA-approved regions

Gibco™ fetal bovine sera is specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.

$1,466.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin Countries meeting USDA importation requirements
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12662029
Encompass
View Documents Promotion Details
Gibco™
12662029
500 mL
Each for $1,466.00
Only null left
 
Description

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Origin: Countries meeting USDA importation requirements
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Storage: Store at -5°C to -20°C.

Specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Fetal
Liquid
Fetal Bovine Serum
Sterile
≤25 mg/dL
Frozen
Cattle/Bovine
Countries meeting USDA importation requirements
Bottle
Standard (Sterile-filtered)
<50 EU/mL
MS Cell-Qualified
Videos
SDS
Documents

Documents

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.