Learn More
Gibco™ Fetal Bovine Serum, Premium
Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.
Supplier: Gibco™ A5670602
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Sera category: Premium
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco serum delivers
- ISO 13485–certified; processed in FDA registered facilities
- Triple-filtered at 0.1 μm
- Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Chemical Identifiers
| Fetal | |
| Frozen | |
| Triple-filtered, 0.1 μm | |
| Premium |
| Refer to CoA for specific origin* | |
| Fetal Bovine Serum | |
| *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. |
Specifications
| Fetal | |
| Frozen | |
| Fetal Bovine Serum | |
| Gamma-irradiated | |
| Triple-filtered, 0.1 μm | |
| No | |
| USDA Safety Testing Performed for Mexican and Australian origins |
| Refer to CoA for specific origin* | |
| One Shot | |
| 10 x 50 mL | |
| Sterile | |
| *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. | |
| Premium | |
| Yes |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.