missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Premium
GREENER_CHOICE

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.

Supplier:  Gibco™ A5670401

 View more versions of this product

Catalog No. A5670401


Only null left

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

Specifications

Chemical Identifiers

Fetal
Frozen
Triple-filtered, 0.1 μm
Premium
Refer to CoA for specific origin*
Fetal Bovine Serum
*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

Specifications

Store at ≤-10°C.
Fetal
Frozen
Fetal Bovine Serum
Sterile-filtered
Premium
≤10 EU/mL
No
Premium
USDA Safety Testing Performed for Mexican and Australian origins
Cattle/Bovine
Refer to CoA for specific origin*
One Shot
50 mL
Triple-filtered, 0.1 μm
Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance
*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
≤25 mg/dL
Dry Ice
No
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 5 Years

SDS
Documents

Documents

Promotions

Promotions

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.