missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, dialyzed, US origin
GREENER_CHOICE

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

Supplier:  Gibco™ 26400044

Encompass

 View more versions of this product

Catalog No. 26400044


Only null left

Description

Description

Gibco Fetal bovine sera include a broad range of specialty sera, earning the trust of researchers with consistent quality and award-winning support that helps meet your unique research requirements. Dialyzed FBS provides a more controlled and defined environment with the removal of small molecules, enhancing outcome accuracy and reliability.

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
Specifications

Chemical Identifiers

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Fetal Bovine Serum
Frozen
Fetal
Liquid
Dialyzed

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Fetal
Liquid
Fetal Bovine Serum
Dialyzed
Cattle/Bovine
United States
Bottle
500 mL
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.