missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals FerroZine™ Iron Reagent, Hydrate, 95+%, Pure

Catalog No. AC410570010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410570010 1 g Glass bottle
AC410570050 5 g Glass bottle
AC410570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC410570010 Supplier Thermo Scientific Chemicals Supplier No. 410570010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1266615-85-3 | C20H13N4NaO6S2 | 492.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1266615-85-3
Molecular Formula C20H13N4NaO6S2
Molecular Weight (g/mol) 492.46
MDL Number MFCD00150794
InChI Key ZGVNYCXXBQPDPQ-UHFFFAOYSA-M
Synonym ferrozine mono-sodium salt hydrate, ferrozine™ iron reagent, c20h13n4o6s2.na.h2o, pdt disulfonate monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p inverted exclamation marka-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid monosodium salt hydrate
PubChem CID 129893581
IUPAC Name sodium 4-[3-(pyridin-2-yl)-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzene-1-sulfonate
SMILES [Na+].OS(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color Yellow
Physical Form Crystalline Powder
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95+%
Solubility Information Solubility in water: soluble
Formula Weight 492.46
Percent Purity ≥95%
Grade Pure
Chemical Name or Material FerroZine* iron reagent,hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only