missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018864
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018864 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018864 Supplier Merck Emd Millipore Supplier No. PHR14831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ferrous sulfate is the sulfate salt of divalent cation form of iron. It is an essential body mineral, that is commonly used in the treatment of iron deficiency anemia.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 7782-63-0
Vapor Pressure 14.6 mmHg (25°C)
Density 1.898 g/mL (at 25°C)
Linear Formula FeSO4 · 7H2O
MDL Number MFCD00149719
Synonym Iron(II) sulfate heptahydrate; Ferrous sulfate heptahydrate
Quantity 1 g
Formula Weight 278.01
Grade Certified Reference Material