missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate, Purified (Iron (II) Sulfate Heptahydrate), Reagents

Catalog No. p-200020880
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337242 2.5 kg
01337241 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337242 Supplier Labchem Inc Supplier No. C22070002.5A5
Only null left

Ferrous Sulfate Heptahydrate, Purified (Iron (II) Sulfate Heptahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym Iron(II) Sulfate
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >300°C
Color Green or Bluish-Green
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.897
Chemical Name or Material Ferrous Sulfate Heptahydrate, Purified (Iron (II) Sulfate Heptahydrate)