missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate Heptahydrate MP Biomedicals
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Formula Weight | 278 |
| Percent Purity | 99% |
| Density | 1.9g/mL |
| Chemical Name or Material | Ferrous Sulfate, Heptahydrate |