missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, Heptahydrate, Granular, USP, 99.5-104.5%, Spectrum™ Chemical

Catalog No. p-7157472
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
2.5 kg
50 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18602723 500 g Poly Bottle
18602724 2.5 kg Poly Pail
18602725 12 kg Poly Pail
18602726 50 kg Fiber Drum
4 options

Catalog No. 18602723

Supplier: Spectrum Chemical Mfg Cor FE110500GM

Only null left

FE110, 7782-63-0, FeSO4·7H2O

Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Granular, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 99.5 to 104.5%
Grade USP
Packaging Poly Bottle