missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate, Heptahydrate, Granular, USP, 99.5-104.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Granular, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 12 kg |
| Percent Purity | 99.5 to 104.5% |
| Grade | USP |
| Packaging | Poly Pail |