missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate (Crystalline), ACS Grade, Fisher Chemical™

Catalog No. p-32113
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I146500 500 g Poly Bottle
I1463 3 kg Poly Bottle
I14610 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I146500 Supplier Thermo Fisher Scientific Supplier No. I146 500
Only null left

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 300°C
Melting Point 64°C
Color Blue Green
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.0%
Formula Weight 278.01
Percent Purity ≥99.0%
Grade ACS Grade
Identification Pass test
Packaging Poly Bottle
Decomposition Information >300°C
Density 1.898g/cm³
Chemical Name or Material Ferrous Sulfate Heptahydrate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:1