missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate Heptahydrate (Crystalline), ACS Grade, Fisher Chemical™
Specifications
Specifications
| Boiling Point | 300°C |
| Melting Point | 64°C |
| Color | Blue Green |
| Physical Form | Solid |
| Quantity | 3 kg |
| Assay Percent Range | ≥99.0 % |
| Formula Weight | 278.01 |
| Percent Purity | ≥99.0% |
| Grade | Certified ACS |
| Identification | Pass test |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Harmful if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:1 Instability:1 |